Merck
  • Acetaminophen glucuronide and plasma glucose report identical estimates of gluconeogenesis and glycogenolysis for healthy and prediabetic subjects using the deuterated water method.
Acetaminophen glucuronide and plasma glucose report identical estimates of gluconeogenesis and glycogenolysis for healthy and prediabetic subjects using the deuterated water method.
Magnetic resonance in medicine (2012-10-02)
Cristina Barosa, John G Jones, Robert Rizza, Ananda Basu, Rita Basu
RESUMO
Plasma glucose (2) H-enrichment in positions 5 ((2) H5) and 2 ((2) H2) from deuterated water ((2) H2 O) provides a measure of the gluconeogenic contribution to endogenous glucose production. Urinary glucuronide analysis can circumvent blood sampling but it is not known if glucuronide and glucose enrichments are equal. Thirteen subjects with impaired fasting glucose/impaired glucose tolerance and 11 subjects with normal fasting glucose and normal glucose tolerance ingested (2) H2 O to ∼0.5% body water and acetaminophen. Glucose and glucuronide (2) H5 and (2) H2 were measured by (2) H NMR spectroscopy of monoacetone glucose. For normal fasting glucose/normal glucose tolerance, (2) H5 was 0.23 ± 0.02% and 0.25 ± 0.02% for glucose and glucuronide, respectively, whereas (2) H2 was 0.47 ± 0.01% and 0.49 ± 0.02%, respectively. For impaired fasting glucose/impaired glucose tolerance, (2) H5 was 0.22 ± 0.01% and 0.26 ± 0.02% for glucose and glucuronide, respectively, whereas (2) H2 was 0.46 ± 0.01% and 0.49 ± 0.02%, respectively. The gluconeogenic contribution to endogenous glucose production measured from glucose and glucuronide were identical for both normal fasting glucose/normal glucose tolerance (48 ± 4 vs. 51 ± 3%) and impaired fasting glucose/impaired glucose tolerance (48 ± 2 vs. 53 ± 3%).
MATERIAIS
Número do produto
Marca
Descrição do produto
Supelco
Paracetamol β-D-glucuronide, analytical standard