Merck
  • A highly sensitive LC-MS/MS method capable of simultaneously quantitating celiprolol and atenolol in human plasma for a cassette cold-microdosing study.
A highly sensitive LC-MS/MS method capable of simultaneously quantitating celiprolol and atenolol in human plasma for a cassette cold-microdosing study.
Journal of separation science (2011-05-20)
Yoshiyuki Minamide, Yukio Osawa, Hiroshi Nishida, Harue Igarashi, Shinobu Kudoh
RESUMO
A highly sensitive simultaneous quantitative method for a cassette cold-microdosing study on celiprolol and atenolol was developed with liquid chromatography-tandem mass spectrometry. The method utilizes a combination of solid-phase extraction (SPE) with strong cation exchange (SCX) cartridge columns and reversed-phase chromatography with an ODS analytical column. SCX-SPE cartridge columns (100 mg sorbent) were used for a selective extraction of celiprolol, atenolol and metoprolol (internal standard) from 500 μL of human plasma samples. Turbo-ion spray at positive mode was employed for the ionization of the drug compounds. Quantitation was performed on a triple quadrupole mass spectrometer by selected reaction monitoring with the transitions of m/z 380 to m/z 251 for celiprolol and m/z 267 to m/z 145 for atenolol. Separation of analytes was achieved on an ODS column (100 mm length × 2.1 mm id, 3 μm) by a gradient elution with 10 mM formic acid and methanol by varying their proportion at a flow rate of 0.2 mL/min. The method was validated in the range of 1-250 pg/mL for celiprolol and 2.5-250 pg/mL for atenolol and was successfully applied to the elucidation of pharmacokinetic profiling in a cold cassette microdosing study of the β-blockers.