Merck
[Determination of acetylharpagide in Ajuga decumbens by HPLC].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2006-05-19)
Dang-Kun Yue, Guang-Zhi Ding, Wei-Hao Wang, Zhi-Min Wang, Jian-Rong Li, Yan Tong
RESUMO
To establish a method of the quantitative determination of acetylharpagide in Ajuga decumbens. The chromatographic conditions were as follows: a Phenomenex Luna C18 column was used, the mobile phase was composed of acetontrile-water (15:85), the flow rate was 1.0 mL x min(-1) and the UV absorbance detection was set at 197 nm. Linearity of acetylharpagide was in the range of 0.6-3.6 microg (r = 0.9993), and the average recovery and RSD were 99.13% and 2.48%, respectively. The contents of acetylharpagide ranged 0.40%-6.39% in A. decumbens. The method was simple, accurate and sensitive.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
8-Acetylharpagide, ≥85% (LC/MS-ELSD)