Merck
Essential role of Ca2+/calmodulin in Early Endosome Antigen-1 localization.
Molecular biology of the cell (2003-07-15)
Deirdre C Lawe, Nachida Sitouah, Susan Hayes, Anil Chawla, Joseph V Virbasius, Richard Tuft, Kevin Fogarty, Lawrence Lifshitz, David Lambright, Silvia Corvera
RESUMO
Ca2+ is an essential requirement in membrane fusion, acting through binding proteins such as calmodulin (CaM). Ca2+/CaM is required for early endosome fusion in vitro, however, the molecular basis for this requirement is unknown. An additional requirement for endosome fusion is the protein Early Endosome Antigen 1 (EEA1), and its recruitment to the endosome depends on phosphatidylinositol 3-phosphate [PI(3)P] and the Rab5 GTPase. Herein, we demonstrate that inhibition of Ca2+/CaM, by using either chemical inhibitors or specific antibodies directed to CaM, results in a profound inhibition of EEA1 binding to endosomal membranes both in live cells and in vitro. The concentration of Ca2+/CaM inhibitors required for a full dissociation of EEA1 from endosomal membranes had no effect on the activity of phosphatidylinositol 3-kinases or on endogenous levels of PI(3)P. However, the interaction of EEA1 with liposomes containing PI(3)P was decreased by Ca2+/CaM inhibitors. Thus, Ca2+/CaM seems to be required for the stable interaction of EEA1 with endosomal PI(3)P, perhaps by directly or indirectly stabilizing the quaternary organization of the C-terminal FYVE domain of EEA1. This requirement is likely to underlie at least in part the essential role of Ca2+/CaM in endosome fusion.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
N-(6-Aminohexyl)-1-naphthalenesulfonamide hydrochloride, ≥98% (HPLC)