Merck
  • Suzuki-Miyaura cross-coupling of potassium dioxolanylethyltrifluoroborate and aryl/heteroaryl chlorides.
Suzuki-Miyaura cross-coupling of potassium dioxolanylethyltrifluoroborate and aryl/heteroaryl chlorides.
Organic letters (2013-03-16)
Nicolas Fleury-Brégeot, Daniel Oehlrich, Frederik Rombouts, Gary A Molander
RESUMO
A robust and efficient protocol for the introduction of the dioxolanylethyl moiety onto various aryl and heteroaryl halides has been developed, providing cross-coupling yields up to 93%. Copper-catalyzed borylation of 2-(2-bromoethyl)-1,3-dioxolane with bis(pinacolato)diboron followed by treatment with potassium bifluoride provides the key organotrifluoroborate reagent.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
2-(2-Bromoethyl)-1,3-dioxolane, 96%