Merck
  • Glucagon-like peptide-1 receptor imaging for the localisation of insulinomas: a prospective multicentre imaging study.
Glucagon-like peptide-1 receptor imaging for the localisation of insulinomas: a prospective multicentre imaging study.
The lancet. Diabetes & endocrinology (2014-03-14)
Emanuel Christ, Damian Wild, Susanne Ederer, Martin Béhé, Guillaume Nicolas, Martyn E Caplin, Michael Brändle, Thomas Clerici, Stefan Fischli, Christoph Stettler, Peter J Ell, Jochen Seufert, Beat Gloor, Aurel Perren, Jean Claude Reubi, Flavio Forrer
RESUMO
Small benign insulinomas are hard to localise, leading to difficulties in planning of surgical interventions. We aimed to prospectively assess the insulinoma detection rate of single-photon emission CT in combination with CT (SPECT/CT) with a glucagon-like peptide-1 receptor avid radiotracer, and compare detection rates with conventional CT/MRI techniques. In our prospective imaging study, we enrolled adults aged 25-81 years at centres in Germany, Switzerland, and the UK. Eligible patients had proven clinical and biochemical endogenous hyperinsulinaemic hypoglycaemia and no evidence for metastatic disease on conventional imaging. CT/MRI imaging was done at referring centres according to standard protocols. At three tertiary nuclear medicine centres, we used whole body planar images and SPECT/CT of the abdomen up to 168 h after injection of (111)In-[Lys40(Ahx-DTPA-(111)In)NH2]-exendin-4 ((111)In-DTPA-exendin-4) to identify insulinomas. Consenting patients underwent surgery and imaging findings were confirmed histologically. Between Oct 1, 2008, and Dec 31, 2011, we recruited 30 patients. All patients underwent (111)In-DTPA-exendin-4 imaging, 25 patients underwent surgery (with histological analysis), and 27 patients were assessed with CT/MRI. (111)In-DTPA-exendin-4 SPECT/CT correctly detected 19 insulinomas and four additional positive lesions (two islet-cell hyperplasia and two uncharacterised lesions) resulting in a positive predictive value of 83% (95% CI 62-94). One true negative (islet-cell hyperplasia) and one false negative (malignant insulinoma) result was identified in separate patients by (111)In-DTPA-exendin-4 SPECT/CT. Seven patients (23%) were referred to surgery on the basis of (111)In-DTPA-exendin-4 imaging alone. For 23 assessable patients, (111)In-DTPA-exendin-4 SPECT/CT had a higher sensitivity (95% [95% CI 74-100]) than did CT/MRI (47% [27-68]; p=0.011). (111)In-DTPA-exendin-4 SPECT/CT could provide a good second-line imaging strategy for patients with negative results on initial imaging with CT/MRI. Oncosuisse, the Swiss National Science Foundation, and UK Department of Health.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Glycocholic acid hydrate, synthetic, ≥97% (HPLC)