Merck
Gait speed in Parkinson disease correlates with cholinergic degeneration.
Neurology (2013-10-01)
Nicolaas I Bohnen, Kirk A Frey, Stephanie Studenski, Vikas Kotagal, Robert A Koeppe, Peter J H Scott, Roger L Albin, Martijn L T M Müller
RESUMO
We investigated dopaminergic and cholinergic correlates of gait speed in Parkinson disease (PD) and non-PD control subjects to test the hypothesis that gait dysfunction in PD may result from multisystem degeneration. This was a cross-sectional study. Subjects with PD but without dementia (n = 125, age 65.6 ± 7.3 years) and elderly subjects without PD (n = 32, age 66.0 ± 10.6 years) underwent [¹¹C]dihydrotetrabenazine dopaminergic and [(11)C]methyl-4-piperidinyl propionate acetylcholinesterase PET imaging, and cognitive and clinical testing, including an 8.5-m walk in the dopaminergic "off" state. The fifth percentile of cortical cholinergic activity in the elderly without PD was used to define normal-range activity in the subjects with PD. Normal-range cortical cholinergic activity was present in 87 subjects with PD (69.6%). Analysis of covariance using gait speed as the dependent variable demonstrated a significant model (F = 6.70, p < 0.0001) with a significant group effect (F = 3.36, p = 0.037) and significant slower gait speed in the low cholinergic PD subgroup (0.97 ± 0.22 m/s) with no significant difference between the normal-range cholinergic PD subgroup (1.12 ± 0.20 m/s) and control subjects (1.17 ± 0.18 m/s). Covariate effects were significant for cognition (F = 6.58, p = 0.011), but not for striatal dopaminergic innervation, sex, or age. Comorbid cortical cholinergic denervation is a more robust marker of slowing of gait in PD than nigrostriatal denervation alone. Gait speed is not significantly slower than normal in subjects with PD with relatively isolated nigrostriatal denervation.