Merck
Angelicin regulates LPS-induced inflammation via inhibiting MAPK/NF-κB pathways.
The Journal of surgical research (2013-07-03)
Fang Liu, Guo-quan Sun, Hua-yi Gao, Rui-sheng Li, Lanan-Wassy Soromou, Na Chen, Yan-hong Deng, Hai-hua Feng
RESUMO
Angelicin is a furocoumarin found in Psoralea corylifolia L. fruit. The purpose of this study was to investigate the protective ability of angelicin against inflammation in lipopolysaccharide (LPS)-stimulated RAW 264.7 cells and LPS-induced in vivo acute lung injury model. The concentrations of tumor necrosis factor alpha (TNF-α) and interleukin (IL)-6 in the culture supernatants of RAW 264.7 cells were determined 24 h after LPS administration. ALI was induced by intratracheal instillation of LPS. Six hours after LPS inhalation, bronchoalveolar lavage fluid and lung tissue samples were obtained for enzyme-linked immunosorbent assay, histologic, and Western blotting analyses. The results showed that pretreatment with angelicin markedly downregulated TNF-α and IL-6 levels in vitro and in vivo, and significantly decreased the amount of inflammatory cells, lung wet-to-dry weight ratio, and myeloperoxidase activity in LPS-induced ALI mice. Furthermore, Western blotting analysis results demonstrated that angelicin blocked the phosphorylation of IκBα, NF-κBp65, p38 MAPK, and JNK in LPS-induced ALI. These results suggest that angelicin was potentially advantageous to prevent inflammatory diseases by inhibiting NF-κB and MAPK pathways. Our data indicated that angelicin might be a potential new agent for prevention of inflammatory reactions and diseases in the clinic.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Angelicin