Merck
Formation of perfluorinated surfactants from precursors by indigenous microorganisms in groundwater.
Chemosphere (2013-06-12)
Michio Murakami, Hiroshi Nishikoori, Hiroshi Sakai, Kumiko Oguma, Hideshige Takada, Satoshi Takizawa
RESUMO
The formation of perfluorinated surfactants (PFSs) from their precursors in waters is of concern. In this study, the formation of PFSs through biodegradation of precursors was measured in incubation tests. Indigenous microorganisms in groundwater were able to biodegrade perfluorooctane sulfonamide (FOSA) to yield perfluorooctane sulfonate (PFOS). The addition of nutrients and soil promoted the formation. A 42-d incubation test using sources of groundwater recharge showed that PFOS, perfluorooctanoate, and perfluorononanoate were significantly and remarkably (⩾1.5×) formed from precursors in street runoff through biodegradation, but not in rainwater or wastewater effluent. Significant formation of PFSs from precursors in street runoff was observed.
MATERIAIS
Número do produto
Marca
Descrição do produto
Supelco
Heptadecafluorooctanesulfonic acid solution, 100 μg/mL in methanol, analytical standard
Preço e disponibilidade não estão disponíveis no momento.