Merck
Regulation of mitochondrial morphogenesis by annexin A6.
PloS one (2013-01-24)
Marcin Chlystun, Michelangelo Campanella, Ah-Lai Law, Michael R Duchen, Lux Fatimathas, Tim P Levine, Volker Gerke, Stephen E Moss
RESUMO
Mitochondrial homeostasis is critical in meeting cellular energy demands, shaping calcium signals and determining susceptibility to apoptosis. Here we report a role for anxA6 in the regulation of mitochondrial morphogenesis, and show that in cells lacking anxA6 mitochondria are fragmented, respiration is impaired and mitochondrial membrane potential is reduced. In fibroblasts from AnxA6(-/-) mice, mitochondrial Ca(2+) uptake is reduced and cytosolic Ca(2+) transients are elevated. These observations led us to investigate possible interactions between anxA6 and proteins with roles in mitochondrial fusion and fission. We found that anxA6 associates with Drp1 and that mitochondrial fragmentation in AnxA6(-/-) fibroblasts was prevented by the Drp1 inhibitor mdivi-1. In normal cells elevation of intracellular Ca(2+) disrupted the interaction between anxA6 and Drp1, displacing anxA6 to the plasma membrane and promoting mitochondrial fission. Our results suggest that anxA6 inhibits Drp1 activity, and that Ca(2+)-binding to anxA6 relieves this inhibition to permit Drp1-mediated mitochondrial fission.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Anti-ANXA6 (AB2) antibody produced in rabbit, IgG fraction of antiserum