Merck
  • Gas-phase reactivity of the O=P(OCH3)2 + phosphonium ion towards alpha,beta-unsaturated esters in a quadrupole ion trap.
Gas-phase reactivity of the O=P(OCH3)2 + phosphonium ion towards alpha,beta-unsaturated esters in a quadrupole ion trap.
Journal of mass spectrometry : JMS (2005-02-16)
Eric Leclerc, Emmanuelle Leon, Marie-Hélène Taphanel, Jean-Pierre Morizur
RESUMO
Ion-molecule reactions between the O=P(OCH3)2 + phosphonium ions and eight alpha,beta-unsaturated esters (methyl acrylate, ethyl acrylate, methyl crotonate, ethyl crotonate, methyl 3,3-dimethylacrylate, ethyl 3,3-dimethylacrylate, methyl methacrylate and ethyl methacrylate) were performed in a quadrupole ion trap mass spectrometer. The O=P(OCH3)2 + phosphonium ions, formed by electron ionization from neutral trimethyl phosphite, were found to react with alpha,beta-unsaturated esters to give an adduct [RR'C=CR''COOR''', O=P(OCH3)2]+, which lose spontaneously a molecule of trimethyl phosphate (R'''=CH3) or dimethyl ethyl phosphate (R'''=C2H5). An ion corresponding to a protonated trialkyl phosphate is also observed when substituent R''=H. To confirm the experimental results, and to elucidate the mechanism for the formation of the ionic species, a theoretical study using the density functional theory (DFT) approach was carried out. The potential energy surface obtained from B3LYP/6-31G(d,p) calculations for the reaction between O=P(OCH3)2 + and methyl acrylate is described.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Ethyl 3,3-dimethylacrylate, 98%