Merck
  • Synthesis of Novel Functionalized 4-Aza-2,3-Didehydropodophyllotoxin Derivatives with Potential Antitumor Activity.
Synthesis of Novel Functionalized 4-Aza-2,3-Didehydropodophyllotoxin Derivatives with Potential Antitumor Activity.
Journal of heterocyclic chemistry (2011-01-05)
Ajay Kumar, Antonio E Alegria
RESUMO
Novel arylamino alcohols were synthesized and these alcohols were used to prepare 12 novel N-(2-hydroxy-ethyl)-2,3-didehydroazapodophyllotoxins, in one step, by simple reflux in ethanol. Isolated yields in the range of 50-70% were obtained.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
1,4-Benzodioxan-6-amine, ≥98%