Merck
  • One-pot cyclization of 2-aminophenethyl alcohols: a novel and direct approach to the synthesis of N-acyl indolines.
One-pot cyclization of 2-aminophenethyl alcohols: a novel and direct approach to the synthesis of N-acyl indolines.
The Journal of organic chemistry (2007-11-02)
Zengxue Wang, Wen Wan, Haizhen Jiang, Jian Hao
RESUMO
A unique one-pot cyclization of 2-aminophenethyl alcohols with carboxylic acids in the presence of PPh3, CCl4, and NEt3 furnished the formation of N-acyl indolines in good to excellent yields. This new approach provides an efficient, scalable, low-cost, and direct access to the biologically important indolines which are further oxidizable to indoles and oxindoles.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
2-Aminophenethyl alcohol, 97%