Merck
  • Cyanide modulated fluorescent supramolecular assembly of a hexaphenylbenzene derivative for detection of trinitrotoluene at the attogram level.
Cyanide modulated fluorescent supramolecular assembly of a hexaphenylbenzene derivative for detection of trinitrotoluene at the attogram level.
Chemical communications (Cambridge, England) (2012-12-19)
Vandana Bhalla, Subhamay Pramanik, Manoj Kumar
RESUMO
Fluorescent nanorods formed by self-assembly of hexaphenylbenzene derivative in the presence of cyanide ions serve as a sensitive colorimetric and fluorogenic sensor for the detection of trinitrotoluene (TNT) at the attogram (10(-18) g) level with a detection limit of 10.21 ppq (parts per quadrillion).