Merck
The biology of oxygen.
The European respiratory journal (2008-04-02)
P D Wagner
RESUMO
The biology of O(2) is extremely complex and defies a comprehensive yet brief summary that showcases its roles across the entire animal and plant kingdom. This necessarily short introduction to the 2006 Taormina Lung Science Conference (Taormina, Italy) on hypoxia will examine key features of the biology of O(2) only within mammalian cells, even that being a daunting task.