Merck
  • The influence of reaction conditions on the Diels-Alder cycloadditions of 2-thio-3-chloroacrylamides; investigation of thermal, catalytic and microwave conditions.
The influence of reaction conditions on the Diels-Alder cycloadditions of 2-thio-3-chloroacrylamides; investigation of thermal, catalytic and microwave conditions.
Organic & biomolecular chemistry (2010-10-12)
Marie Kissane, Denis Lynch, Jay Chopra, Simon E Lawrence, Anita R Maguire
RESUMO
The Diels-Alder cycloadditions of cyclopentadiene and 2,3-dimethyl-1,3-butadiene to a range of 2-thio-3-chloroacrylamides under thermal, catalytic and microwave conditions is described. The influence of reaction conditions on the outcome of the cycloadditions, in particular the stereoselectivity and reaction efficiency, is discussed. While the cycloadditions have been attempted at the sulfide, sulfoxide and sulfone levels of oxidation, use of the sulfoxide derivatives is clearly beneficial for stereoselective construction of Diels-Alder cycloadducts.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
2,3-Dimethyl-1,3-butadiene, 98%, contains 100 ppm BHT as stabilizer