Merck
Introducing manganese complexes as redox mediators for dye-sensitized solar cells.
Physical chemistry chemical physics : PCCP (2014-01-08)
Ishanie Rangeeka Perera, Akhil Gupta, Wanchun Xiang, Torben Daeneke, Udo Bach, Richard A Evans, C André Ohlin, Leone Spiccia
RESUMO
The abundance and low toxicity of manganese have led us to explore the application of manganese complexes as redox mediators for dye sensitized solar cells (DSCs), a promising solar energy conversion technology which mimics some of the key processes in photosynthesis during its operation. In this paper, we report the development of a DSC electrolyte based on the tris(acetylacetonato)manganese(iii)/(iv), [Mn(acac)3](0/1+), redox couple. PEDOT-coated FTO glass was used as a counter electrode instead of the conventionally used platinum. The influence of a number of device parameters on the DSC performance was studied, including the concentration of the reduced and oxidized mediator species, the concentration of specific additives (4-tert-butylpyridine, lithium tetrafluoroborate, and chenodeoxycholic acid) and the thickness of the TiO2 working electrode. These studies were carried out with a new donor-π-acceptor sensitizer K4. Maximum energy conversion efficiencies of 3.8% at simulated one Sun irradiation (AM 1.5 G; 1000 W m(-2)) with an open circuit voltage (VOC) of 765 mV, a short-circuit current (JSC) of 7.8 mA cm(-2) and a fill factor (FF) of 0.72 were obtained. Application of the commercially available MK2 and N719 sensitizers resulted in an energy conversion efficiency of 4.4% with a VOC of 733 mV and a JSC of 8.6 mA cm(-2) for MK2 and a VOC of 771 mV and a JSC of 7.9 mA cm(-2) for N719. Both dyes exhibit higher incident photon to current conversion efficiencies (IPCEs) than K4.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
4-tert-Butylpyridine, 98%