Merck
ATM binds to beta-adaptin in cytoplasmic vesicles.
Proceedings of the National Academy of Sciences of the United States of America (1998-08-26)
D S Lim, D G Kirsch, C E Canman, J H Ahn, Y Ziv, L S Newman, R B Darnell, Y Shiloh, M B Kastan
RESUMO
Inherited mutations in the ATM gene lead to a complex clinical phenotype characterized by neuronal degeneration, oculocutaneous telangiectasias, immune dysfunction, and cancer predisposition. Using the yeast two-hybrid system, we demonstrate that ataxia telangiectasia mutated (ATM) binds to beta-adaptin, one of the components of the AP-2 adaptor complex, which is involved in clathrin-mediated endocytosis of receptors. The interaction between ATM and beta-adaptin was confirmed in vitro, and coimmunoprecipitation and colocalization studies show that the proteins also associate in vivo. ATM also interacts in vitro with beta-NAP, a neuronal-specific beta-adaptin homolog that was identified as an autoantigen in a patient with cerebellar degeneration. Our data describing the association of ATM with beta-adaptin in vesicles indicate that ATM may play a role in intracellular vesicle and/or protein transport mechanisms.