Merck
Azelaic acid--biochemistry and metabolism.
Acta dermato-venereologica. Supplementum (1989-01-01)
S Passi, M Picardo, G Mingrone, A S Breathnach, M Nazzaro-Porro
RESUMO
Medium chain length dicarboxylic acids (DA) from C8 to C13 are competitive inhibitors of tyrosinase in vitro. The introduction of electron acceptor groups or electron donor groups into the 2 and/or the 8 position of the molecule enhances or reduces respectively the inhibitory effects of DA. In addition to tyrosinase, DA can reversibly inhibit thioredoxin reductase, NADPH cytochrome P450 reductase, NADH dehydrogenase, succinic dehydrogenase and H2CoQ-Cytochrome C oxidoreductase. Among DA, azelaic acid (AA, C9 dicarboxylic acid) is extensively used because: 1) it is much cheaper than other DA; 2) it has no apparent toxic or teratogenic or mutagenic effect; 3) when administered perorally to humans, at the same concentrations as the other DA, it reaches much higher serum and urinary concentrations. Serum concentrations and urinary excretion obtained with intravenous or intra-arterial infusions of AA are significantly higher than those achievable by oral administration. Together with AA, variable amounts of its catabolites, mainly pimelic acid, are found in serum and urine, indicating an involvement of mitochondrial beta-oxidative enzymes. Short-lived serum levels of AA follow a single 1 h intravenous infusion, but prolonging the period of infusion with successive doses of similar concentration produces sustained higher levels during the period of administration. These levels are consistent with the concentrations of AA capable of producing a cytotoxic effect on tumoral cells in vitro. AA is capable of crossing the blood-brain barrier: its concentration in the cerebrospinal fluid is normally in the range of 2-5% of the values in the serum.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Pimelic acid, 98%