Merck
  • The metalloprotease disintegrin ADAM8. Processing by autocatalysis is required for proteolytic activity and cell adhesion.
The metalloprotease disintegrin ADAM8. Processing by autocatalysis is required for proteolytic activity and cell adhesion.
The Journal of biological chemistry (2002-10-10)
Uwe Schlomann, Dirk Wildeboer, Ailsa Webster, Olga Antropova, Dagmar Zeuschner, C Graham Knight, Andrew J P Docherty, Marc Lambert, Lisa Skelton, Harald Jockusch, Jörg W Bartsch
RESUMO
ADAMs (a disintegrin and metalloprotease domains) are metalloprotease and disintegrin domain-containing transmembrane glycoproteins with proteolytic, cell adhesion, cell fusion, and cell signaling properties. ADAM8 was originally cloned from monocytic cells, and its distinct expression pattern indicates possible roles in both immunology and neuropathology. Here we describe our analysis of its biochemical properties. In transfected COS-7 cells, ADAM8 is localized to the plasma membrane and processed into two forms derived either by prodomain removal or as remnant protein comprising the extracellular region with the disintegrin domain at the N terminus. Proteolytic removal of the ADAM8 propeptide was completely blocked in mutant ADAM8 with a Glu(330) to Gln exchange (EQ-A8) in the Zn(2+) binding motif (HE(330)LGHNLGMSHD), arguing for autocatalytic prodomain removal. In co-transfection experiments, the ectodomain but not the entire MP domain of ADAM8 was able to remove the prodomain from EQ-ADAM8. With cells expressing ADAM8, cell adhesion to a substrate-bound recombinant ADAM8 disintegrin/Cys-rich domain was observed in the absence of serum, blocked by an antibody directed against the ADAM8 disintegrin domain. Soluble ADAM8 protease, consisting of either the metalloprotease domain or the complete ectodomain, cleaved myelin basic protein and a fluorogenic peptide substrate, and was inhibited by batimastat (BB-94, IC(50) approximately 50 nm) but not by recombinant tissue inhibitor of matrix metalloproteinases 1, 2, 3, and 4. Our findings demonstrate that ADAM8 processing by autocatalysis leads to a potential sheddase and to a form of ADAM8 with a function in cell adhesion.