Merck
Paroxetine efficacy in the treatment of generalized anxiety disorder.
Acta psychiatrica Scandinavica (1997-05-01)
P Rocca, V Fonzo, M Scotta, E Zanalda, L Ravizza
RESUMO
Recently, there has been a renewed interest in alternatives to the benzodiazepines for the treatment of generalized anxiety disorder (GAD). The aim of the present study was to compare the efficacy of paroxetine vs. imipramine and 2'-chlordesmethyldiazepam in 81 patients with a DSM-IV diagnosis of GAD. Approximately two-thirds of the patients who completed the study improved greatly or moderately on all three active drugs. During the first 2 weeks of treatment, 2'-chlordesmethyldiazepam treatment resulted in the greatest improvement in anxiety ratings. Both paroxetine and imipramine treatment resulted in more improvement than 2'-chlordesmethyldiazepam by the fourth week of treatment. Paroxetine and imipramine affect predominantly psychic symptoms, whereas 2'-chlordesmethyldiazepam affects predominantly somatic symptoms. Our results suggest that paroxetine is effective for the treatment of GAD.