Merck
Anti-inflammatory activity of 2,3-dihydrophthalazine-1,4-diones in CF1 mice.
International journal of tissue reactions (1996-01-01)
L Butner, Y Huang, E Tse, I H Hall
RESUMO
2,3-Dihydrophthalazine-1,4-diones were observed to be potent anti-inflammatory agents as well as capable of protecting against endotoxin shock in mice at 8 mg/kg i.p. These agents blocked both locally- and centrally-induced pain at 8 mg/kg i.p. In part they appear to mediate their effects due to their ability to suppress the release of cytokines such as TNF alpha and IL-1 from macrophages, as well as the binding of these cytokines to high-affinity receptors on target cells involved in the inflammation process. Lysosomal hydrolytic enzymes as well as 5'-lipoxygenase activities were reduced by the agents.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Phthalhydrazide, ReagentPlus®, 99%