Merck
  • PEG thiazolidine-2-thione, a novel reagent for facile protein modification: conjugation of bovine hemoglobin.
PEG thiazolidine-2-thione, a novel reagent for facile protein modification: conjugation of bovine hemoglobin.
Bioconjugate chemistry (1996-11-01)
R B Greenwald, A Pendri, A Martinez, C Gilbert, P Bradley
RESUMO
A novel PEG linker that employs a thiazolidine-2-thione group has been synthesized. Kinetic studies done on this compound demonstrate a relatively long half-life compared to those of traditional succinimidyl linkers. This new PEG derivative reacts with proteins under mild conditions and was utilized to conjugate bovine hemoglobin (bHb) to provide a PEG amide-linked protein. The physical characteristics of this conjugate were compared with those of the known PEG carbamate-linked bHb.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
2-Thiazoline-2-thiol, 98%