Merck
Genetic manipulation of the pathway for diacetyl metabolism in Lactococcus lactis.
Applied and environmental microbiology (1996-07-01)
S R Swindell, K H Benson, H G Griffin, P Renault, S D Ehrlich, M J Gasson
RESUMO
Diacetyl is an important food flavor compound produced by certain strains of citrate-metabolizing lactic acid bacteria. Citrate is converted to pyruvate, from which diacetyl is produced via intermediate alpha-acetolactate. This paper reports the cloning and analysis of the gene (aldB) encoding alpha-acetolactate decarboxylase from Lactococcus lactis MG1363. Deletion of the MG1363 chromosomal aldB gene was achieved by double crossover homologous recombination. The mutant strain was found to produce diacetyl; alpha-acetolactate decarboxylase activity was eliminated. Overexpression of the cloned ilvBN genes (encoding an alpha-acetolactate synthase) in the aldB deletion strain produced even higher levels of alpha-acetolactate, acetoin, and diacetyl.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
(±)-2-Acetoxypropionic acid, ≥97.0% (GC)