Merck
Determination of rhoifolin and daidzin in human plasma by high-performance liquid chromatography.
Journal of chromatography. B, Biomedical applications (1994-05-13)
K Ishii, S Urano, T Furuta, Y Kasuya
RESUMO
A method for determining flavonoids in human plasma is presented for application to pharmacokinetic studies of two flavonoids, rhoifolin and daidzin. Isocratic reversed-phase high-performance liquid chromatography (HPLC) was used with genistin as an internal standard and solid-phase extraction using a Sep-Pak C18 cartridge. The mobile phases were acetonitrile-0.1 M ammonium acetate solution (20:80, v/v) for rhoifolin and methanol-0.1 M ammonium acetate solution (33:67, v/v) for daidzin. The detection limits on-column were 2 ng for rhoifolin and 0.5 ng for daidzin.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Apigenin 7-O-neohesperidoside, ≥99.0% (HPLC)