- Histological investigations of the antiarthrotic effect of tribenoside (Glyvenol) in experimental osteoarthrosis.
Histological investigations of the antiarthrotic effect of tribenoside (Glyvenol) in experimental osteoarthrosis.
The Journal of rheumatology (1983-04-01)
D A Kalbhen, E Scherbach, K Felten
PMID6345770
RESUMO
Experiments in adult hens with chemically induced osteoarthrosis or osteoarthritis (OA) of the knee joint have shown that daily treatment with tribenoside (Glyvenol) in single oral doses of 50 mg/kg or 150 mg/kg markedly reduced the intensity and frequency of joint degeneration. We investigated the inhibitory effect of tribenoside on OA by histological techniques. Comparisons of articular cartilage from untreated and treated animals showed no qualitative differences in the alterations of the morphology and histopathology of the tribenoside treated specimens. Only the frequency and intensity of degenerative alterations were markedly reduced in the drug treated animals.
MATERIAIS
Número do produto
Marca
Descrição do produto
Tribenoside, European Pharmacopoeia (EP) Reference Standard