Merck
  • [The enzymatic degradation of DDT. 6. The fate of the DDT metabolites dichlorodiphenyldichloroethylene and dichlorodiphenylmonochloroethylene in the rat].
[The enzymatic degradation of DDT. 6. The fate of the DDT metabolites dichlorodiphenyldichloroethylene and dichlorodiphenylmonochloroethylene in the rat].
Die Nahrung (1987-01-01)
M Kujawa, R Macholz, R Knoll, R Plass
RESUMO
DDE is hydrogenated in the rat organism. The distribution of the compound is described. DDMU is degradated to DBP as well as transformed to DDA and DDOH. Excretion and distribution are studied.