Merck
  • Effects of serotonin1 and serotonin2 receptor agonists and antagonists on blood pressure, heart rate and sympathetic nerve activity.
Effects of serotonin1 and serotonin2 receptor agonists and antagonists on blood pressure, heart rate and sympathetic nerve activity.
The Journal of pharmacology and experimental therapeutics (1987-09-01)
R B McCall, B N Patel, L T Harris
RESUMO
Previous studies indicate that serotonin (5-HT) neurons provide a tonic excitatory input to central sympathetic neurons. The purpose of the present study was to utilize a number of 5-HT agonists in order to provide insights into the general function of the serotonergic system in the regulation of central sympathetic pathways. The 5-HT1A agonists 8-hydroxy-dipropylaminotetralin and p-aminophenyl-ethyl-m-trifluoromethylphenyl piperazine produced a dose-related inhibition of sympathetic nerve discharge (SND) recorded from either the postganglionic inferior cardiac nerve or the preganglionic splanchnic nerve in chloralose-anesthetized cats. The sympatholytic effects of 8-hydroxy-dipropylaminotetralin and p-aminophenyl-ethyl-m-trifluoromethylphenyl piperazine were accompanied by hypotension and bradycardia. The effects of 5-HT1A agonists were reversed by the 5-HT1A antagonist spiperone. In contrast, spiperone alone produced decreases in blood pressure, heart rate and SND. The 5-HT1B agonists 1[3(trifluoromethyl) phenyl]-piperazine, 1-(3-chlorophenyl) piperazine and 1-(2-methoxyphenyl) piperazine all produced variable effects on SND. In some experiments, SND was increased by these agents, whereas it was decreased in others. The 5-HT2 agonist 1-(2,5-dimethoxy-4-iodophenyl)-2-aminopropane produced a marked increase in SND. A second 5-HT2 agonist, MK212, produced similar effects. The role of 5-HT receptor subtypes in mediating the 5-HT excitation of sympathetic neurons is discussed. It is suggested that 5-HT1A agonists inhibit SND through a process of disfacilitation by inhibiting the firing rate of 5-HT neurons. Possible mechanisms by which 5-HT2 agonists increase SND are proposed.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
LY-165,163, solid