Merck
  • Effects of nicotinic acid, nicotinamide, and pyridylcarbinol in pharmacological dosages on lipid metabolism in humans.
Effects of nicotinic acid, nicotinamide, and pyridylcarbinol in pharmacological dosages on lipid metabolism in humans.
International journal for vitamin and nutrition research. Supplement = Internationale Zeitschrift fur Vitamin- und Ernahrungsforschung. Supplement (1989-01-01)
N Zöllner
RESUMO
The clinical use of nicotinic acid, nicotinamide, and pyridylcarbinol as drugs against hypercholesterolemia is critically reviewed. Though several questions remain open as to the mode of action of these compounds, it is concluded that they indeed belong to the most useful drugs for the treatment of hypercholesterolemia and for the prevention of coronary disease.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
3-Pyridinemethanol, 98%