Merck
  • Removal of carbamazepine and clofibric acid from water using double templates-molecularly imprinted polymers.
Removal of carbamazepine and clofibric acid from water using double templates-molecularly imprinted polymers.
Environmental science and pollution research international (2013-02-26)
Chao-meng Dai, Juan Zhang, Ya-lei Zhang, Xue-fei Zhou, Yan-ping Duan, Shu-guang Liu
RESUMO
A novel double templates-molecularly imprinted polymer (MIP) was prepared by precipitation polymerization using carbamazepine (CBZ) and clofibric acid (CA) as the double templates molecular and 2-vinylpyridine as functional monomer. The equilibrium data of MIP was well described by the Freundlich isotherm model. Two kinetic models were adopted to describe the experimental data, and the pseudo second-order model well-described adsorption of CBZ and CA on the MIP. Adsorption experimental results showed that the MIP had good selectivity and adsorption capacity for CBZ and CA in the presence of competitive compounds compared with non-imprinted polymer, commercial powdered activated carbon, and C18 adsorbents. The feasibility of removing CBZ and CA from water by the MIP was demonstrated using tap water, lake water, and river water.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
2-(p-Chlorophenoxy)-2-methylpropionic acid, 97%