Merck
  • Ginsenoside Rd attenuates mitochondrial dysfunction and sequential apoptosis after transient focal ischemia.
Ginsenoside Rd attenuates mitochondrial dysfunction and sequential apoptosis after transient focal ischemia.
Neuroscience (2011-01-12)
R Ye, X Zhang, X Kong, J Han, Q Yang, Y Zhang, Y Chen, P Li, J Liu, M Shi, L Xiong, G Zhao
RESUMO
We previously found that ginsenoside Rd (Rd), one of the major active ingredients in Panax ginseng, protects neuronal cells from hydrogen peroxide and oxygen-glucose deprivation, an in vitro model of cerebral ischemia. In this study, we examined the protective effects of Rd in an animal model of focal cerebral ischemia. Rats administered with Rd or vehicle were subjected to transient middle cerebral artery occlusion (MCAO). Rd (50 mg/kg) significantly reduced the infarct volume by 52.8%. This reduction of injury volume was associated with an improvement in neurological function and was sustained for at least 2 weeks after the induction of ischemia. To evaluate the underlying mechanisms of Rd against stroke, brain tissues were assayed for mitochondrial enzyme activities, mitochondrial membrane potential (MMP), production of reactive oxygen species (ROS), energy metabolites, and apoptosis. Rd markedly protected mitochondria as indicated by preserved respiratory chain complex activities and aconitase activity, lowered mitochondrial hydrogen peroxide production, and hyperpolarized MMP. Microdialysis results illustrated that Rd significantly decreased the accumulation of lactate, the end product of anaerobic glycolysis, and increased pyruvate, the end product of aerobic glycolysis, hence inducing a lower lactate/pyruvate ratio. Additionally, in vitro studies further exhibited that Rd protected isolated mitochondria from calcium-induced damage by attenuating mitochondrial swelling, preserving MMP and decreasing ROS production. Moreover, Rd treatment reduced mitochondrial release of cytochrome c (CytoC) and apoptosis-inducing factor (AIF), thereby minimizing mitochondria-mediated apoptosis following ischemia. In conclusion, these findings demonstrated that Rd exerts neuroprotective effects in transient focal ischemia, which may involve an integrated process of the mitochondrial protection, energy restoration and inhibition of apoptosis.
MATERIAIS
Número do produto
Marca
Descrição do produto
Supelco
Ginsenoside Rd, analytical standard