Merck
[Speed drugs].
Przeglad lekarski (2009-01-01)
Wojciech Piekoszewski, Ewa Florek
RESUMO
Until recently "speed drugs" have not awake higher social emotions. Usually they associate with Red Bull, allowed driving long time, and workaholic from modern enterprise to regenerate before next task. Currently on the base of new regulation on the list of controlled substances benzylpyperazine, its related compounds and 17 plants products were added. The article discussed the sources and biological action of these psychoactive substances, which are illegal in Poland.