Merck
Evaluation of the kappa-opioid receptor-selective tracer [(11)C]GR103545 in awake rhesus macaques.
European journal of nuclear medicine and molecular imaging (2010-02-17)
Bent W Schoultz, Trine Hjornevik, Frode Willoch, János Marton, Akihiro Noda, Yoshihiro Murakami, Sosuke Miyoshi, Shintaro Nishimura, Erik Arstad, Alexander Drzezga, Ichiro Matsunari, Gjermund Henriksen
RESUMO
The recent development in radiosynthesis of the (11)C-carbamate function increases the potential of [(11)C]GR103545, which for the last decade has been regarded as promising for imaging the kappa-opioid receptor (kappa-OR) with PET. In the present study, [(11)C]GR103545 was evaluated in awake rhesus macaques. Separate investigations were performed to clarify the OR subtype selectivity of this compound. Regional brain uptake kinetics of [(11)C]GR103545 was studied 0-120 min after injection. The binding affinity and opioid subtype selectivity of [(11)C]GR103545 was determined in cells transfected with cloned human opioid receptors. In vitro binding assays demonstrated a high affinity of GR103545 for kappa-OR (K(i) = 0.02 +/- 0.01 nM) with excellent selectivity over mu-OR (6 x 10(2)-fold) and) delta-OR (2 x 10(4)-fold). PET imaging revealed a volume of distribution (V(T)) pattern consistent with the known distribution of kappa-OR, with striatum = temporal cortex > cingulate cortex > frontal cortex > parietal cortex > thalamus > cerebellum. [(11)C]GR103545 is selective for kappa-OR and holds promise for use to selectively depict and quantify this receptor in humans by means of PET.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
GR 89696 fumarate salt, solid