- Chirality change of chloronitrobenzene on Au(111) induced by inelastic electron tunneling.
Chirality change of chloronitrobenzene on Au(111) induced by inelastic electron tunneling.
Angewandte Chemie (International ed. in English) (2009-04-01)
Violeta Simic-Milosevic, Jörg Meyer, Karina Morgenstern
PMID19334019
RESUMO
Chirality chameleon: Inelastic electron tunneling manipulation can be used to change a single chloronitrobenzene (ClNB) molecule, randomly adsorbed on Au(111), into its desired enantiomeric form (r or l, see STM images and ball-and-stick representation) and to vary its rotational orientation. The different threshold voltages for chirality change (260 mV) and rotation (380 mV) allow these processes to be induced separately.