Merck
  • Synthesis, primary photophysical and antibacterial properties of naphthyl ester cinoxacin and nalidixic acid derivatives.
Synthesis, primary photophysical and antibacterial properties of naphthyl ester cinoxacin and nalidixic acid derivatives.
Journal of photochemistry and photobiology. B, Biology (2008-06-20)
Franklin Vargas, Tamara Zoltan, Carlos Rivas, Alvaro Ramirez, Tulynan Cordero, Yrene Díaz, Carla Izzo, Ylec M Cárdenas, Verónica López, Lubimar Gómez, Jessenia Ortega, Alberto Fuentes
RESUMO
We have synthesized two naphthyl ester quinolone derivates and determined their ability to generate reactive oxygen species (ROS) such as (1)O(2), ()OH, H(2)O(2) upon photolysis with UV-A light. The ability of cinoxacin (1) and nalidixic acid (2), and their naphthyl ester derivatives (3 and 4) to generate a dose-dependent amount of singlet oxygen and ROS (()(-)O(2), ()OH) in cell-free systems was detected by histidine assay and by luminol-enhanced chemiluminescence (LCL), respectively. Their electronic absorption and emission spectra were quantified and their photostability was determined. Their tendency to generate peroxidic derivative species showed the following order: 3>4; in contrast, their ability to generate singlet oxygen was 4>3 and these were better sensitizers than their parent quinolones 1 and 2. The antibacterial activity in darkness and under irradiation of compounds 3 and 4 was tested on Escherichia coli and compared with that of their parent compounds. An enhanced antibacterial activity by irradiation of the naphthyl esters of cinoxacin and nalidixic acid on E. coli was observed.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Cinoxacin