Merck
  • The effect of intrahippocampal injection of group II and III metobotropic glutamate receptor agonists on anxiety; the role of neuropeptide Y.
The effect of intrahippocampal injection of group II and III metobotropic glutamate receptor agonists on anxiety; the role of neuropeptide Y.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology (2006-11-30)
Maria Smiałowska, Joanna M Wierońska, Helena Domin, Barbara Zieba
RESUMO
Earlier studies conducted by our group and by other authors indicated that metabotropic glutamatergic receptor (mGluR) ligands might have anxiolytic activity and that amygdalar neuropeptide Y (NPY) neurons were engaged in that effect. Apart from the amygdala, the hippocampus, another limbic structure, also seems to be engaged in regulation of anxiety. It is rich in mGluRs and contains numerous NPY interneurons. In the present study, we investigated the anxiolytic activity of group II and III mGluR agonists after injection into the hippocampus, and attempted to establish whether hippocampal NPY neurons and receptors were engaged in the observed effects. Male Wistar rats were bilaterally microinjected with the group II mGluR agonist (2S,1'S,2'S)-2-(carboxycyclopropyl)glycine (L-CCG-I), group III mGluR agonist O-Phospho-L-serine (L-SOP), NPY, the Y1 receptor antagonist BIBO 3304, and the Y2 receptor antagonist BIIE 0246 into the CA1 or dentate area (DG). The effect of those compounds on anxiety was tested in the elevated plus-maze. Moreover, the effects of L-CCG-I and L-SOP on the expression of NPYmRNA in the hippocampus were studied using in situ hybridization method. It was found that a significant anxiolytic effect was induced by L-SOP injection into the CA1 region or by L-CCG-I injection into the DG. The former effect was inhibited by BIBO 3304, the latter by BIIE 0246. NPY itself showed antianxiety action after injection into both structures. In the CA1 area, the effect of NPY was prevented by BIBO 3304, whereas in the DG by BIIE 0246. Both the mGluR agonists L-CCG-I and L-SOP induced a potent increase in NPYmRNA expression in the DG region of the hippocampal formation. The obtained results indicate that group II and III mGluR agonists, L-CCG-I and L-SOP, as well as NPY display anxiolytic activity in the hippocampus, but act differently in the CA1 and DG. It was observed that group III mGluRs and Y1 receptors were engaged in the response in the CA1 area, whereas group II mGluRs and Y2 receptors played a pivotal role in the DG region.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
(2S,3R,4S)-α-(Carboxycyclopropyl)glycine, solid