Merck
Analysis of wave packet motion in frequency and time domain: oxazine 1.
The journal of physical chemistry. A (2006-08-11)
Markus Braun, Constanze Sobotta, Regina Dürr, Horst Pulvermacher, Stephan Malkmus
RESUMO
Wave packet motion in the laser dye oxazine 1 in methanol is investigated by spectrally resolved transient absorption spectroscopy. The spectral range of 600-690 nm was accessible by amplified broadband probe pulses covering the overlap region of ground-state bleach and stimulated emission signal. The influence of vibrational wave packets on the optical signal is analyzed in the frequency domain and the time domain. For the analysis in the frequency domain an algorithm is presented that accounts for interference effects of neighbored vibrational modes. By this method amplitude, phase and decay time of vibrational modes are retrieved as a function of probe wavelength and distortions due to neighbored modes are reduced. The analysis of the data in the time domain yields complementary information on the intensity, central wavelength, and spectral width of the optical bleach spectrum due to wave packet motion.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Basic Blue 3, Dye content 25 %