Merck
  • Laminin-10/11 and fibronectin differentially prevent apoptosis induced by serum removal via phosphatidylinositol 3-kinase/Akt- and MEK1/ERK-dependent pathways.
Laminin-10/11 and fibronectin differentially prevent apoptosis induced by serum removal via phosphatidylinositol 3-kinase/Akt- and MEK1/ERK-dependent pathways.
The Journal of biological chemistry (2002-03-14)
Jianguo Gu, Akemi Fujibayashi, Kenneth M Yamada, Kiyotoshi Sekiguchi
RESUMO
Cell adhesion to the extracellular matrix inhibits apoptosis, but the molecular mechanisms underlying the signals transduced by different matrix components are not well understood. Here, we examined integrin-mediated antiapoptotic signals from laminin-10/11 in comparison with those from fibronectin, the best characterized extracellular adhesive ligand. We found that the activation of protein kinase B/Akt in cells adhering to laminin-10/11 can rescue cell apoptosis induced by serum removal. Consistent with this, wortmannin, a specific inhibitor of phosphatidylinositol 3-kinase, or ectopic expression of a dominant-negative mutant of Akt selectively accelerated cell death upon serum removal. In contrast to laminin-10/11, fibronectin rescued cells from serum depletion-induced apoptosis mainly through the extracellular signal-regulated kinase pathway. Cell survival on fibronectin but not laminin was significantly reduced by treatment with PD98059, a specific inhibitor of mitogen- or extracellular signal-regulated kinase kinase-1 (MEK1) and by expression of a dominant-negative mutant of MEK1. Laminin-10/11 was more potent than fibronectin in preventing apoptosis induced by serum depletion. These results, taken together, demonstrate laminin-10/11 potency as a survival factor and demonstrate that different extracellular matrix components can transduce distinct survival signals through preferential activation of subsets of multiple integrin-mediated signaling pathways.