Merck
  • Dithiolane analogs of lignans inhibit interferon-gamma and lipopolysaccharide-induced nitric oxide production in macrophages.
Dithiolane analogs of lignans inhibit interferon-gamma and lipopolysaccharide-induced nitric oxide production in macrophages.
Acta pharmacologica Sinica (2001-08-15)
I M Hussaini, Y H Zhang, J J Lysiak, T Y Shen
RESUMO
To investigate the effect of a group of novel synthetic dithiolane analogs of lignans and a well characterized platelet-activating factor (PAF) receptor antagonist, L659,989 on PAF-receptor binding, IFN-gamma- and lipopolysaccharide (LPS)-induced NO production, and steady-state inducible nitric-oxide synthase (iNOS) mRNA expression. PAF-receptor binding study was performed by displacement of 3H-PAF from rabbit platelet membrane; NO production was quantitated by measuring the NO oxidation product, nitrite, in conditioned culture medium; expression of iNOS mRNA was assessed by Northern blot analysis. The dithiolane analogs inhibited the production of NO, decreased iNOS mRNA expression and antagonized PAF-receptor binding. L659,989 had no effect on NO production and iNOS mRNA expression. Among the compounds tested, there was no simple correlation between their PAF-receptor antagonistic and iNOS inhibitory activities. The dithiolane analogs are a new synthetic chemical class of iNOS expression regulators with dual biologic functions: inhibiting iNOS induction and blocking PAF-receptor.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
1,3-Dithiolane, 97%