Merck
  • The reaction of thiolates with 2,3-dibromo-1-propanol revisited: application to the synthesis of bis(fattyalkylthio)propanols.
The reaction of thiolates with 2,3-dibromo-1-propanol revisited: application to the synthesis of bis(fattyalkylthio)propanols.
Chemistry and physics of lipids (1999-10-26)
M D Gernon, J G Turcotte, R H Notter, M Mu, Y Lear, J L Snelgrove, A L Schwan
RESUMO
This work compares two reaction schemes for preparing 2,3-bis(fattyalkylthio)-1-propanols for further synthetic adaptation as hydrophobic analogs of lung surfactant phosphatidylcholines. An attempt to prepare 2,3-bis(fattyalkylthio)-1-propanols based on the previously published methods of Bell and co-workers (B.R. Ganong, C.R. Loomis, Y.A. Hannun, R.M. Bell, 1986. Proc. Natl. Acad. Sci. USA 83, 1184-1188; B.R. Ganong, R.M. Bell, 1987. Methods Enzymol. 141, 313-320; J.P. Walsh, L. Fahrner, R.M. Bell, 1990. J. Biol. Chem. 265, 4374-4381) was found to give the rearranged 1,3-bis(fattyalkylthio)-2-propanols as major products. As a reliable alternative, the reaction of ethyl 2,3-dibromopropionate with 2 equivalents of long chain sodium n-alkanethioates gave the corresponding ethyl 2,3-bis(n-alkylthio)propionates, which were then reduced with LiAlH4 to yield the desired 2,3-bis(fattyalkylthio)-1-propanols. Both 13C and 1H NMR spectroscopy were used to differentiate the two possible 1,3- and 2,3-dithio substituted alcohol products and to rigorously assign their structures.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
2,3-Dibromo-1-propanol, 98%