Merck
  • Profiling of amine metabolites in human biofluids by micellar electrokinetic chromatography with laser-induced fluorescence detection.
Profiling of amine metabolites in human biofluids by micellar electrokinetic chromatography with laser-induced fluorescence detection.
Analytical and bioanalytical chemistry (2007-03-21)
Hua-Ming Tseng, Yin Li, David A Barrett
RESUMO
A rapid and sensitive capillary electrophoresis (CE) method has been developed for profiling organic metabolites containing amine functional groups in mammalian biofluids. Metabolites containing an amine group were derivatized with 4-fluoro-7-nitrobenzo-2,1,3-oxadiazol (NBD-F), separated by micellar electrokinetic chromatography (MEKC), and detected by argon-ion (488 nm) laser-induced fluorescence detection. The optimized MEKC background electrolyte conditions were: 50 mmol L-1 sodium cholate, 5 mmol L-1 beta-cyclodextrin, and 20 mmol L-1 Brij 35 in 20 mmol L-1 aqueous borate buffer, pH 9.3, containing 7% methanol. Under these conditions all the amine compounds in mammalian biofluids, for example plasma, saliva, and urine, were derivatized directly, without extraction, in a minimum volume of 100 nL and the derivatives could be separated within 16 min. Up to 90% of the amine-containing metabolites in plasma and saliva could be identified by reference to standard compounds. For twenty amine standards linearity of calibration was better than R2=0.99. Migration-time and peak-area reproducibility were better than RSD 1.5% and 15% respectively. In replicate analysis of human plasma bioanalytical precision ranged between 0.7 and 3.8 RSD% for a 5.0-microL volume and between 1.7 and 5.5 RSD% for 100-nL volume. The concentrations measured were found to be in agreement with literature values.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
4-Fluoro-7-nitrobenzofurazan, ≥98% (elemental analysis)
Sigma-Aldrich
4-Fluoro-7-nitrobenzofurazan, BioReagent, suitable for fluorescence, ≥98.0% (HPLC)