Merck
Efficacy of decoquinate against Neospora caninum tachyzoites in cell cultures.
Veterinary parasitology (1997-01-01)
D S Lindsay, J M Butler, B L Blagburn
RESUMO
Neospora caninum is a major cause of abortion in dairy cattle in the United States and other countries. Abortions and neonatal mortality also occur in other ruminant species. Decoquinate is an anticoccidial that is approved for use in cattle and goats in the United States. We studied the efficacy of decoquinate against tachyzoites of N. caninum in a 5-day of treatment, cell culture flask lesion-based assay. Decoquinate killed tachyzoites at concentrations of 0.1 and 0.01 microgram ml-1. Decoquinate had little measurable effect on extracellular tachyzoites. Decoquinate acted quickly to kill intracellular stages at coccidiocidal concentrations; tachyzoites were killed within 5 min at 0.1 microgram ml-1 decoquinate.
MATERIAIS
Número do produto
Marca
Descrição do produto
Supelco
Decoquinate, VETRANAL®, analytical standard