- Isolation of guaianoid pigments from the gorgonian Calicogorgia granulosa.
Isolation of guaianoid pigments from the gorgonian Calicogorgia granulosa.
Journal of natural products (1996-10-01)
Y Seo, J R Rho, N Geum, J B Yoon, J Shin
PMID8984163
RESUMO
Three pigments of the guaiazulene class have been isolated from the gorgonian Calicogorgia granulosa. Structures of these compounds have been determined as guaiazulene, 2,2'- diguaiazulenylmethane, and a new compound, 2,2'-biguaiazulenyl, by combined chemical and spectroscopic methods. 2,2'-Biguaiazulenyl exhibited moderate antimicrobial activity.
MATERIAIS