- The effect of the composition of food on the absorption of sulfameter.
The effect of the composition of food on the absorption of sulfameter.
International journal of clinical pharmacology and biopharmacy (1979-06-01)
S Kaumeier
PMID468450
RESUMO
The present investigation is concerned with the influence of the composition of food on the absorption of sulfameter. Six physically healthy patients each were given 2 g of sulfameter simultaneously with a high lipid, high protein and high carbohydrate test meal. This experiment was designed as a threefold crossover study, and there was a randomized assignment of patients to the different conditions. The results show that sulfameter is significantly better absorbed when administered with a high lipid meal than when given with a high protein or high carbohydrate meal, demonstrated by the areas under the serum concentration curves (AUC), by the peak serum concentration and by the cumulative renal excretion.
MATERIAIS