Merck
Digitalis-induced augmentation of cardiopulmonary baroreflex control of forearm vascular resistance.
Circulation (1985-01-01)
T Imamura, A Takeshita, T Ashihara, K Yamamoto, S Hoka, M Nakamura
RESUMO
We sought to determine whether digitalis augments cardiopulmonary baroreflex control of forearm vascular resistance in normal young men. Cardiopulmonary baroreceptor input was reduced with lower body negative pressure (LBNP) at 10 and 20 mm Hg, which decreased central venous pressure but did not alter blood pressure or heart rate. Decreases in forearm blood flow and increases in forearm vascular resistance with LBNP were greater after administration of lanatoside C (Cedilanid) than before, and the slope of the regression line relating changes in central venous pressure and those in forearm vascular resistance was steeper after lanatoside C. Vasoconstrictor responses to the cold pressor test did not differ before and after lanatoside C, which suggested that augmented responses to LBNP after the drug were not caused by a generalized change in reflex control. These results suggest that lanatoside C augments the tonic inhibitory influence of cardiopulmonary baroreceptors in normal men.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Lanatoside C, ≥97% (TLC)