Merck
Diphacinone-induced coagulopathy in the dog.
Journal of the American Veterinary Medical Association (1986-02-15)
A Schulman, R Lusk, C L Lippincott, S J Ettinger
RESUMO
Five cases of coagulopathy caused by consumption of indanedione (diphacinone)-based rodenticides are reported. In each case, acute onset of lethargy and respiratory distress were the predominant initial clinical signs. Thoracic radiography revealed pulmonary edema, pleural effusion, and/or pericardial effusion as consistent findings. Laboratory evaluations confirmed coagulopathies that responded to vitamin K1 therapy.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
1,3-Indandione, 97%