- Some observations on the nature of the calcium-cresolphthalein complexone reaction and its relevance to the clinical laboratory.
Some observations on the nature of the calcium-cresolphthalein complexone reaction and its relevance to the clinical laboratory.
Annals of clinical biochemistry (1987-07-01)
C M Corns, C J Ludman
PMID3662386
RESUMO
The reasons for the non-linearity of the standard curve for the calcium-cresolphthalein complexone reaction have been investigated. Calcium forms both 1:1 and 2:1 complexes with cresolphthalein complexone; at low calcium concentrations the 1:1 complex predominates and causes non-linearity. At high calcium concentrations the cresolphthalein complexone concentration becomes limiting, resulting in flattening of the calibration curve. It is recommended that two-point calibration of the assay be performed using a low calcium standard rather than setting on the baseline.
MATERIAIS
Número do produto
Marca
Descrição do produto
Preço e disponibilidade não estão disponíveis no momento.