- Hexamminecobaltic chloride provides a simple method for marking neurones for electron microscopy.
Hexamminecobaltic chloride provides a simple method for marking neurones for electron microscopy.
Brain research (1988-03-29)
P M Littlewood, P J Simmons
PMID3365553
RESUMO
Intracellular injection of hexamminecobaltic chloride into a neurone causes characteristic changes to the structure of its mitochondria. This provides a simple technique to label neuronal profiles for examination of their ultrastructure. Synaptic structures are not affected by hexamminecobaltic ions.