Merck
  • 25-hydroxyvitamin D₃ and 1,25-dihydroxyvitamin D₃ promote the differentiation of human subcutaneous preadipocytes.
25-hydroxyvitamin D₃ and 1,25-dihydroxyvitamin D₃ promote the differentiation of human subcutaneous preadipocytes.
PloS one (2012-12-29)
Hataikarn Nimitphong, Michael F Holick, Susan K Fried, Mi-Jeong Lee
RESUMO
1,25(OH)(2)D(3) inhibits adipogenesis in mouse 3T3-L1 adipocytes, but little is known about its effects or local metabolism in human adipose tissue. We showed that vitamin D receptor (VDR) and 1α-hydroxylase (CYP27B1), the enzyme that activates 25(OH)D(3) to 1,25(OH)(2)D(3), were expressed in human adipose tissues, primary preadipocytes and newly-differentiated adipocytes. Preadipocytes and newly-differentiated adipocytes were responsive to 1,25(OH)(2)D(3), as indicated by a markedly increased expression of CYP24A1, a primary VDR target. 1,25(OH)(2)D(3) enhanced adipogenesis as determined by increased expression of adipogenic markers and triglyceride accumulation (50% to 150%). The magnitude of the effect was greater in the absence of thiazolidinediones. 1,25(OH)(2)D(3) was equally effective when added after the removal of differentiation cocktail on day 3, but it had no effect when added only during the induction period (day 0-3), suggesting that 1,25(OH)(2)D(3) promoted maturation. 25(OH)D(3) also stimulated CYP24A1 expression and adipogenesis, most likely through its conversion to 1,25(OH)(2)D(3). Consistent with this possibility, incubation of preadipocytes with 25(OH)D(3) led to 1,25(OH)(2)D(3) accumulation in the media. 1,25(OH)(2)D(3) also enhanced adipogenesis in primary mouse preadipocytes. We conclude that vitamin D status may regulate human adipose tissue growth and remodeling.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
3-epi-25-Hydroxyvitamin D3, 98% (CP)