Merck
  • [99mTc(CO)3]+-(HE)3-ZIGF1R:4551, a new Affibody conjugate for visualization of insulin-like growth factor-1 receptor expression in malignant tumours.
[99mTc(CO)3]+-(HE)3-ZIGF1R:4551, a new Affibody conjugate for visualization of insulin-like growth factor-1 receptor expression in malignant tumours.
European journal of nuclear medicine and molecular imaging (2012-11-28)
Anna Orlova, Camilla Hofström, Joanna Strand, Zohreh Varasteh, Mattias Sandstrom, Karl Andersson, Vladimir Tolmachev, Torbjörn Gräslund
RESUMO
Radionuclide imaging of insulin-like growth factor type 1 receptor (IGF-1R) expression in tumours might be used for selection of patients who would benefit from IGF-1R-targeted therapy. We have previously shown the feasibility of IGF-1R imaging using the Affibody molecule (111)In-DOTA-His(6)-Z(IGF1R:4551). The use of (99m)Tc instead of (111)In should improve sensitivity and resolution of imaging, and reduce the dose burden to patients. We hypothesized that inclusion of a HEHEHE tag instead of a His(6) tag in Z(IGF1R:4551) would permit its convenient purification using IMAC, enable labelling with [(99m)Tc(CO)(3)](+), and improve its biodistribution. Z(IGF1R:4551) was expressed with a HEHEHE tag in the N terminus. The resulting (HE)(3)-Z(IGF1R:4551) construct was labelled with [(99m)Tc(CO)(3)](+). Targeting of IGF-1R-expressing cells using [(99m)Tc(CO)(3)](+)-(HE)(3)-Z(IGF1R:4551) was evaluated in vitro and in vivo. (HE)(3)-Z(IGF1R:4551) was stably labelled with (99m)Tc with preserved specific binding to IGF-1R-expressing DU-145 prostate cancer cells in vitro. In mice, [(99m)Tc(CO)(3)](+)-(HE)(3)-Z(IGF1R:4551) accumulated in IGF-1R-expressing organs (pancreas, stomach, lung and salivary gland). [(99m)Tc(CO)(3)](+)-(HE)(3)-Z(IGF1R:4551) demonstrated 3.6-fold lower accumulation in the liver and spleen than (111)In-DOTA-Z(IGF1R:4551). In NMRI nu/nu mice with DU-145 prostate cancer xenografts, the tumour uptake was 1.32 ± 0.11 %ID/g and the tumour-to-blood ratio was 4.4 ± 0.3 at 8 h after injection. The xenografts were visualized using a gamma camera 6 h after injection. (99m)Tc(CO)(3)](+)-(HE)(3)-Z(IGF1R:4551) is a promising candidate for visualization of IGF-1R expression in malignant tumours.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid, ≥97.0% (CHN)